Learn More
Selleck Chemical LLC Amsacrine hydrochloride
SDP
Supplier:  Selleck Chemical LLC S56271G
Encompass_Preferred
Amsacrine hydrochloride (m-AMSA acridinyl anisidide) is the hydrochloride salt form of amsacrine which is an inhibitor of topoisomerase II with antineoplastic activity
Catalog No. 50-000-04013