Learn More
Selleck Chemical LLC AMTB hydrochloride E4673-1G
SDP
Supplier:  Selleck Chemical LLC E46731G
Encompass_Preferred
AMTB hydrochloride is a blocker of the transient receptor potential melastatin 8 (TRPM8) ion channel It reduces the activation of TRPM8 channels induced by icilin as assessed through a Ca2 influx assay with a pIC50 of 6 23 It is used as a tool to evaluate the role of this channel in bladder rhythmic contraction
Catalog No. 50-000-39326