Learn More
Cayman Chemical AmthamInhydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 2950310 MG
Encompass_Preferred
A histamine H2 receptor agonist (pD2 = 6.21 in isolated guinea pig atrium); selective for H2 over H3 receptors (pD2 = 4.7 in isolated guinea pig ileum); increases heart rate in spontaneously beating isolated guinea pig atrium (pD2 = 6.72) and decreases CCK octapeptide-induced contractions in isolated guinea pig gallbladder strips; increases levels of sheep red blood cell-induced production of IgG and IgM antibodies in rabbit serum at 10 UG/kg twice per day
Catalog No. 50-297-8830