Learn More
TARGETMOL CHEMICALS INC Amtolmetin guacil 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1967250MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Amtolmetin guacil (ST-679) inhibits prostaglandin synthesis and cyclooxygenase. Amtolmetin guacil has similar NSAID properties like tolmetin with additional analgesic, antipyretic, and gastroprotective properties. Purity 99.63%
Catalog No. 50-287-6866