Learn More
Medchemexpress LLC Amustaline (dihydrochloride) | 210584-54-6 | MFCD10035286 | 95.0% | 507.28 g·mol⁻1 | C22H27Cl4N3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY106991A10MM 1ML
Encompass_Preferred
Amustaline dihydrochloride is a nucleic acid-targeted alkylator developed for ex vivo pathogen inactivation of red blood cell-containing blood components. Supplied typically as a 10 mM solution in DMSO, it alkylates nucleic acids and breaks down to nonreactive products, enabling inactivation of viruses, bacteria, and protozoa for research and analytical applications.
  • Targets nucleic acids via an acridine intercalator and a bis-alkylator effector.
  • Designed for ex vivo pathogen inactivation of red blood cell-containing components.
  • Supplied as a 10 mM solution in DMSO for small-volume analytical use.
  • Breaks down to nonreactive products through a labile linker hydrolysis.
  • Stored sealed at 4 °C, away from moisture.
Catalog No. 50-002-40474