Learn More
Medchemexpress LLC Amuvatinib (MP470) | 850879-09-3 | MFCD16038298 | 99.4% | 447.5 g/mol | C23H21N5O3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1020650MG
Encompass_Preferred
Amuvatinib is an orally bioavailable, multi-targeted receptor tyrosine kinase inhibitor and DNA repair suppressor used in preclinical research. It inhibits mutant c-Kit, PDGFRα, FLT3, c-Met, and c-Ret, and suppresses RAD51-mediated DNA repair; intended for research use only and not for human or clinical use.
  • Orally bioavailable multi-targeted tyrosine kinase inhibitor.
  • Inhibits mutant c-Kit, PDGFRα, FLT3, c-Met, and c-Ret.
  • Suppresses RAD51-mediated DNA repair, exhibiting antineoplastic activity.
  • Supplied as a research-grade chemical for laboratory applications.
  • Reported molecular formula C23H21N5O3S and molecular weight 447.5 g/mol.
Catalog No. 50-002-45373