Learn More
Medchemexpress LLC Amxi-5001 hydrochloride | 00-00-0 | 98.1% | 493.92 g/mol | C25H21ClFNO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145734A5MG
Encompass_Preferred
AMXI-5001 hydrochloride is a research-use small-molecule hydrochloride salt with dual PARP1/2 inhibitory activity and microtubule polymerization inhibition. It is intended for preclinical biochemical and cellular studies and is supplied with analytical specifications.
  • Dual PARP1/2 and microtubule polymerization inhibitory activity.
  • Suitable for in vitro and in vivo preclinical research.
  • Hydrochloride salt form improves solubility for formulation.
  • Soluble in DMSO at high concentration (≈100 mg/mL) for stock solutions.
  • Solid storage recommended at 4°C; solutions stable at -80°C for extended storage.
  • High chemical purity reported (~98%).
Catalog No. 50-002-21569