Learn More
Cayman Chemical AmycolatopsIn C 100ug
SDP
Supplier:  Cayman Chemical 33255100 UG
Encompass_Preferred
A polyketide macrolide; active against M. bovis and M. tuberculosis (IC50s = 2.7 and 5.7 µM, respectively); cytotoxic to SW620 colorectal and NCI H460 lung cancer cells (IC50s = 10 and 5.9 µM, respectively)
Catalog No. 50-294-7651