Learn More
YMC America, Inc Amyl-SA Actus 5um 150x30mm
SDP
Supplier:  YMC America, Inc KSA99S051530WX
Encompass
YMC s CHIRAL ART series features polysaccharide-derived chiral stationary phases (CSPs) based on amylose and cellulose tris-carbamate derivatives available in both coated and immobilized formats These columns provide exceptional stereoselectivity durability and solvent versatility supporting normal-phase reversed-phase and supercritical fluid chromatography (SFC) modes
Catalog No. 50-008-78187