Learn More
Medchemexpress LLC HY-B2192 500mg Medchemexpress, Amylase CAS:9000-92-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2192500MG
Encompass_Preferred
Medchemexpress, HY-B2192 500mg Amylase CAS:9000-92-4 Amylase is an enzyme produced by pancreas and salivary glands, catalyzing the hydrolysis of starch into sugars. Amylase are broadly classified into α, β, and γ subtypes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2734
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.