Learn More
Medchemexpress LLC Amylin, amide, rat | 124447-81-0 | >99.9% | 3920.44 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP14641MG
Encompass_Preferred
Amylin, amide, rat is a potent, high-affinity ligand for Amylin receptor AMY1 and AMY3 receptors, and variably for AMY2 receptors. This 37 amino acid glucoregulatory hormone, belonging to the calcitonin family of peptides, is synthesized with insulin in the beta cells of pancreatic islets. It regulates glucose homeostasis by inhibiting gastric emptying, suppressing glucagon release, and inducing meal-ending satiety, thereby protecting against postprandial blood glucose spikes and overeating.
  • Potent, high-affinity ligand for Amylin receptor AMY1 and AMY3 receptors
  • Regulates glucose homeostasis
  • Inhibits gastric emptying
  • Suppresses glucagon release
  • Induces meal-ending satiety
  • Protects against postprandial blood glucose spikes and overeating
  • 37 amino acid glucoregulatory hormone
  • Member of the calcitonin family of peptides
Catalog No. 50-004-23110