Learn More
Carbosynth LLC. Amylin (Rat) (0.5 mg vial), 1ea, Disulfide bonds: (Disulfide bond between Cys2 and Cys7), MW: 3920.4, Molecular formula: C167H272N52O53S2, CAS# 124447-81-0
SDP
Supplier:  Carbosynth LLC. PAM4220V0.5MG
Encompass_Preferred
Synonyms: Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-Arg-Ser-Ser-Asn-Asn-Leu-Gly-Pro-Val-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2 APP: Islet Amyloid Polypeptide DAP: Diabetes-Associated Peptide, Storage Temperature: -20°C, References: J.D. Leffert, C.B. Newgard, H. Okamoto, J.L. Milburn, and K.L. Luskey, Proc. Natl. Acad. Sci. USA, 86, 3127 (1989). (Original; cDNA), J. Asai, M. Nakazato, K. Kangawa, S. Matsukura, and H. Mastuo, Biochem. Biophys. Res. Commun., 164, 400 (1989). (Original; Isolation and Structure)
Catalog No. 50-168-6123