Learn More
Medchemexpress LLC Amyloglucosidase, Aspergillus niger | 9032-08-0 | MFCD00081350 | 112.411 | 25 G
Supplier: Medchemexpress LLC 25GHYP2857
Amyloglucosidase, Aspergillus niger is an enzyme derived from many sources, including plants, animals, and microorganisms. It can be used for industrial production and is widely employed for starch saccharification, brewing, and distilling industries.
- Used for starch saccharification, brewing, and distilling
- Derived from Aspergillus niger
- Enzyme activity ≥100000 U/g solid
- Stores at -20°C for 2 years
- For research use only