Learn More
Medchemexpress LLC β-Amyloid (22-35) (TFA) | 144189-71-9 | 96.5% | 1517.64 | 1 MG

Supplier: Medchemexpress LLC HYP1474A1MG
β-Amyloid (22-35) (TFA) is a synthetic peptide, appearing as a white to off-white solid, with a molecular weight of 1517.64. Analyzed by HPLC, it boasts a purity of 96.48%. This product is supplied as a powder and is recommended for research applications.
- White to off-white solid appearance
- High purity of 96.48%
- Molecular weight of 1517.64
- Store as powder sealed and away from moisture
- Powder stable for 2 years at -80°C or 1 year at -20°C
- Compound is unstable in solutions; freshly prepared solutions are recommended
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.