Learn More
Medchemexpress LLC β-Amyloid (25-35), HFIP-treated | 131602-53-4 | 98.2% | 1060.27 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0128A1MG 5
Encompass_Preferred
β-Amyloid (25-35), HFIP-treated is a fragment of the Alzheimer's amyloid β-peptide, demonstrating neurotoxic activities in cultured cells. This product has been treated with HFIP.
  • Fragment of Alzheimer's amyloid β-peptide
  • Demonstrates neurotoxic activities in cultured cells
  • Treated with HFIP
Catalog No. 50-004-26385