Learn More
Bachem Amyloid Beta Protein (1-42) (mouse, rat)
SDP
Supplier:  Bachem 4035885.0500
Encompass_Preferred
Amyloid Beta Protein (1-42) (mouse, rat) , H-Asp-Ala-Glu-Phe-Gly-His-Asp-Ser-Gly-Phe-Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 166090-74-0, C199H307N53O59S, Mr 4418.01, 0.5mg. b-Amyloid Peptide (1-42), mouse, rat. Rodent Ab 1-42 is considerably less aggregation-prone than human Ab 1-42, but antimicrobial activity against a range of clinically relevant microorganisms has been shown for both peptides.
Catalog No. 50-260-012