Learn More
IBL America Amyloid Beta (1-43) (FL) Aβ
Supplier:  IBL America 27710
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human Amyloid Beta (1-43) A in cerebrospinal fluid, brain extract of Tg mouse and cell culture supernatant. For research use only, not for use in diagnostic procedures.This kit can measure full-length (FL) molecules of Human A (1-43).Tetra Methyl Benzidine (TMB) is used as a coloring agent (Chromogen). The strength of coloring is proportional to the quantities of Human A (1-43).
Catalog No. 50-136-6526