Learn More
Sino Biological Peptide Substrates: Amyloid Beta Peptide (1-42)
Catalog No. 502569743
Encompass_Preferred
The Amyloid Beta Peptide (42 aa) sequence is DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA. It is produced synthetically and treated with 1,1,1,3,3,3-Hexafluoro-2-propanol (HFIP) prior to drying which breaks down pre-formed fibrils and monomerizes the peptide.
Catalog No. 50-256-9743 Supplier Sino Biological Supplier No. A4258100
The Amyloid Beta Peptide (42 aa) sequence is DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA. It is produced synthetically and treated with 1,1,1,3,3,3-Hexafluoro-2-propanol (HFIP) prior to drying which breaks down pre-formed fibrils and monomerizes the peptide.