Learn More
CPC Scientific H-Cys-Gly-Ile-Lys-Tyr-Ile-Lys-Asp-Asp-Val-Ile-Leu-Asn-Glu-Pro-Ser-Ala-Asp-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD036A
Encompass
ONE-LETTER SEQUENCE: CGIKYIKDDVILNEPSADMOLECULAR FORMULA: C87H141N21O30S1MOLECULAR WEIGHT:1993.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802078-21-2], SYNONYMS: Amyloid Bri Protein Precursor (89-106)RESEARCH AREA: Alzheimer's Disease
Catalog No. 50-210-8693