Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amyloid P-IN-1 | 1819986-22-5 | 99.59% | 656.68 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY19771100MG

Encompass_Preferred

amyloid P-IN-1 is utilized in research concerning diseases or disorders characterized by the depletion of serum amyloid P component (SAP). These conditions include amyloidosis, Alzheimer's disease, type 2 diabetes mellitus, and osteoarthritis.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 2 years, -20°C for 1 year
  • Solubility (In vitro): Soluble in DMSO (≥ 34 mg/mL or 51.78 mM).

Catalog No. 50-004-01849


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.