Learn More
Medchemexpress LLC amyloid P-IN-1 | 1819986-22-5 | 99.6% | 656.68 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1977110MM/1ML
Encompass_Preferred
amyloid P-IN-1 is a compound utilized in the research of diseases and disorders characterized by the depletion of serum amyloid P component (SAP). This includes conditions such as amyloidosis, Alzheimer's disease, type 2 diabetes mellitus, and osteoarthritis.
  • Formulated for research applications.
  • Solution provided at 10mM concentration in DMSO.
  • High purity for consistent experimental results.
  • Stable under recommended storage conditions for extended periods.
Catalog No. 50-004-00824