Learn More
Medchemexpress LLC Amyloid Precursor Protein Antibody (YA1130) | 100 UL

Supplier: Medchemexpress LLC HYP81385100UL
The Amyloid Precursor Protein Antibody (YA1130) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets human Amyloid Precursor Protein. This antibody is suitable for laboratory research, particularly in immunohistochemistry (paraffin-embedded sections), providing a reliable tool for studying neuroscience applications.
- Monoclonal antibody
- Rabbit host
- Reacts with human protein
- Suitable for immunohistochemistry-paraffin (IHC-P) applications
- Affinity purified
- Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
- Store at -20°C for 1 year
- Avoid repeated freeze/thaw cycles
- Observed molecular weight of 100 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.