Learn More
Medchemexpress LLC Amyloid Precursor Protein Antibody (YA1130) | 50 UL

Supplier: Medchemexpress LLC HYP8138550UL
Amyloid Precursor Protein Antibody (YA1130) is supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide, and 0.05% BSA. This antibody is suitable for IHC-P applications, with a recommended dilution ratio of 1:100-1:200. It is for research use only and has not been fully validated for medical applications.
- Host: Rabbit
- Reactivity: Human
- Application: IHC-P
- Stored at -20°C for 1 year
- Avoid repeated freeze / thaw cycles
- Shipping with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.