Learn More
Newcomer Supply Amyloid, Puchtler Congo Red Stain Kit, 250 ml kit
SDP
Supplier:  Newcomer Supply 9104A
Encompass
Newcomer Supply Amyloid, Puchtler Congo Red Stain Kit procedure is used in identifying the extraneous protein deposits in amyloidosis. The use of polarizing lenses is an essential technique for visualizing amyloid positive areas and/or to confirm negativity.
Catalog No. NC2102257