Learn More
New England Biolabs, Inc. Amylose Magnetic Beads – 25 mg
SDP
Supplier:  New England Biolabs, Inc. E8035S
Encompass
An affinity matrix for the small-scale isolation and purification of maltose binding protein (MBP)-fusion proteins. Amylose is covalently coupled to a paramagnetic particle through a linkage that is stable and leak resistant over a wide pH range.
Catalog No. NC0495571