Learn More
New England Biolabs, Inc. Amylose Resin High Flow 15 ml
SDP
Supplier:  New England Biolabs, Inc. E8022S
Encompass
Amylose Resin High Flow is a cross-linked affinity matrix used for the isolation of proteins fused to maltose-binding protein (MBP). This rigid matrix can be used in automated chromatography systems.
Catalog No. 50-995-071