Learn More
Medchemexpress LLC AN5777 | 491586-42-6 | 98.37% | 319.36 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1616965MG
Encompass_Preferred
AN5777 is a GSPT1 degrader that significantly reduces GSPT1 through the ubiquitin-proteasome system. It induces G1 block and Apoptosis, and also exhibits antitumor activity. It is intended for research use only and not for sale to patients.
  • GSPT1 degrader
  • Significantly reduces GSPT1 through the ubiquitin-proteasome system
  • Induces G1 block and apoptosis
  • Has antitumor activity
Catalog No. 50-003-95360