Learn More
Cayman Chemical AN7 Fatty Acids and Derivative
SDP
Supplier:  Cayman Chemical 1000621210 MG
Encompass_Preferred
A potent antioxidant lipoic acid analog; has 1.5-fold increased maximal capacity over α-lipoic acid to stimulate glucose transport into myocytes
Catalog No. 50-289-0980