Learn More
Apexbio Technology LLC ANA 12 219766-25-3 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B57125.1
Encompass_Preferred
ANA 12 (CAS 219766-25-3) is a potent and selective antagonist of tropomyosin receptor kinase B (TrkB) the receptor for neurotrophins BDNF and NT-4/5 It binds non-competitively to extracellular sites on TrkB exhibiting IC50 values of 45 6 nM (high-affinity site) and 41 1 M (low-affinity site) ANA 12 inhibits BDNF-induced TrkB activation effectively blocking downstream signaling pathways including PLC- PI3K and MAPK In mouse models ANA 12 administration reduces endogenous brain TrkB activity and demonstrates antidepressant and anxiolytic properties supporting its use for researching psychiatric disorders and neuronal signaling
Catalog No. 50-000-62683