Learn More
Medchemexpress LLC Anacardic acid | 16611-84-0 | MFCD07368254 | 98.5% | 348.52 | C22H36O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN202010MG
Encompass_Preferred
Anacardic acid is a natural salicylic acid derivative extracted from cashew nutshell liquid that functions as a histone acetyltransferase (HAT) inhibitor used in biochemical and cell-based epigenetics research. It inhibits HAT activity of p300 and PCAF with reported IC50s of approximately 8.5 μM and 5 μM, respectively.
  • Natural product inhibitor of histone acetyltransferases used for epigenetic studies.
  • High purity solid form suitable for analytical and cell-based assays.
  • Molecular formula C22H36O3 and molecular weight 348.52 g/mol.
  • Available in small solid quantities (e.g., 5 mg, 10 mg, 25 mg) and as a 10 mM solution in DMSO.
  • Stable as powder at -20°C for long-term storage; store solutions at -80°C for extended stability.
Catalog No. 50-002-96523