Learn More
Cayman Chemical Anacardic Acid, 16611-84-0, 5 mg
SDP
Supplier:  Cayman Chemical 131445
Encompass_Preferred
Molecular Formula C22H36O3, Purity ≥98%, Formula Weight 348.5, Synonyms: 2-hydroxy-6-pentadecyl-benzoic acid, 6-pentadecyl Salicylic Acid; An alkyl salicylic acid isolated from cashew shells; inhibits the HAT activity of p300 and PCAF (IC50 = 8.5 and 5 M, respectively); suppresses NF-B activation, inhibits IB phosphorylation, and prohibits p65 nuclear translocation.
Catalog No. NC0775540