Learn More
Medchemexpress LLC HY-12090 5mg Medchemexpress, Anacetrapib CAS:875446-37-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120905MG
Encompass_Preferred
Medchemexpress, HY-12090 5mg Anacetrapib CAS:875446-37-0 Anacetrapib is a potent CETP inhibitor, with IC50s of 7.9±2.5 nM and 11.8±1.9 nM for rhCETP and C13S CETP mutant, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0957