Learn More
Hach Company SC1000 Ethernet Outdoor Kit
Supplier:  Hach Company LZY553
Encompass
SC1000 Ethernet Outdoor Connect KitThis SC1000 Ethernet Outdoor Connect kit includes all of the fittings necessary to adapt the controller s probe module to connect to an ethernet network The user will need to provide the ethernet cable to complete the kit
Catalog No. 50-002-07285