Learn More
Fluotics Analytik Jena Cybio FeliX compatible P50, sterile
Supplier:  Fluotics CYBF60C.F
Encompass
Analytik Jena Cybio FeliX compatible P60 filter/sterile Tips in columns only Working Volume 50uL Tip Color Clear Tip Box R 384 w/ adapter 50 racks/case with 192 tips per rack
Catalog No. 50-004-35178