Learn More
Fluotics Analytik Jena Cybio FeliX compatible P60, non-sterile, Tips in columns only
Supplier:  Fluotics CYBF60C.ST
Encompass
Analytik Jena Cybio FeliX compatible P60 sterile Tips in columns only Working Volume 60uL Tip Color Clear Tip Box R 384 w/ adapter 50 racks/case with 192 tips per rack
Catalog No. 50-004-35134