Learn More
Medchemexpress LLC Anamorelin | 249921-19-5 | 546.70 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1473450MG
Encompass_Preferred
Anamorelin is an orally active Ghrelin receptor agonist that can promote appetite, increase body weight, and stimulate the secretion of growth hormone and insulin-like growth factor-1. It is used in the research of anorexia and cancer cachexia, and is supplied as an analytical standard for research and analytical applications.
  • Orally active ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone and insulin-like growth factor-1 secretion
  • Used in research of anorexia and cancer cachexia
  • Reference standard for qualitative, quantitative, and methodological research experiments
Catalog No. 50-003-61440