Learn More
Medchemexpress LLC Anamorelin | 249921-19-5 | 99.94% | 546.70 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1473410MM/1ML
Encompass_Preferred
Anamorelin (RC-1291) is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. Anamorelin can be used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone secretion
  • Stimulates insulin-like growth factor-1 secretion
  • Used in research of anorexia
  • Used in research of cancer cachexia
Catalog No. 50-003-59981