Learn More
Enzo Life Sciences Anandamide (5mg). CAS: 94421-68-8
SDP
Supplier:  Enzo Life Sciences BMLFA0170005
Encompass_Preferred
Endogenous [1] ligand for the CB1 receptor (CB1: Ki=52nm; CB2: Ki=1930nm [2]) and TRPV1 (Ki=5.78µM [3,4]). Inhibits NF-κB activation through direct binding to IKKβ [5] and induces apoptosis independently of cannabinoid or vanilloid receptors [6]. Activates the MAP kinase (MAPK/ERK) signalling pathway [7]. Alternative name: AEA, N-Arachidonoylethanolamine, (all-Z)-N-(2-Hydroxyethyl)-5,8,11,14-eicosatetraenamide. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (e.g. at a concentration of 50 mg/ml). It is also soluble in DMSO and dimethyl formamide. Long Term Storage: -80°C.
Catalog No. 50-201-0803