Learn More
AdipoGen Andrastin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20144M001
Encompass_Preferred
Chemical. CAS: 174232-42-9. Formula: C28H38O7. MW: 486.6. Isolated from Aspergillus fumigatus. Mycotoxin. Protein farnesyltransferase (PFTase) inhibitor. Anti-cancer compound. Directly interacts with P-glycoprotein and inhibits the efflux of antitumor agents in drug resistant cells.
Catalog No. 50-205-3525