Learn More
Waters Corp Pipetting Robot, Automated Liquid Handling, 0.2 μL to 10 mL Volume Range
Supplier:  Waters Corp 176850100
Encompass_Preferred
The Andrew+ Pipetting Robot is a liquid handling platform designed to streamline and add efficiency to pharmaceutical, life science, and analytical LC-MS workflows. It ensures reproducibility and traceability for laboratory processes and can be used in fully automated antibody purification protocols.
  • Improves workflow efficiency, reproducibility, and traceability.
  • Compatible with common lab hoods and safety cabinets.
  • Connects to Laboratory Information Management Systems (LIMS).
  • Software adapts protocols and simplifies programming.
  • Supports an ecosystem of connected devices.
  • Volume range: 0.2 μL to 10 mL (single-channel) and 0.2 μL to 1.2 mL (eight-channel).
  • Manages up to 11 Dominos per experiment.
Catalog No. NC3951560