Learn More
Genetex Inc Androgen Receptor Monoclonal Antibody [MSVA-367R] HistoMAX™ (GTX04368) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04368
Encompass_Preferred
"The Androgen Receptor antibody [MSVA-367R] HistoMAX™ (GTX04368) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human colonic mucosa tissue using GTX04368 Androgen Receptor antibody [MSVA-367R] HistoMAX. Complete absence of androgen receptor staining in normal colon mucosa."
Catalog No. NC3663023