Learn More
Medchemexpress LLC Androgen receptor Antibody (YA3740) | 99 kDa | 100 UL

Supplier: Medchemexpress LLC HYP84043100UL
Androgen receptor Antibody (YA3740) is a liquid intended for laboratory chemical use and manufacture of substances. It is stable under recommended storage conditions and is not classified as a hazardous substance or mixture.
- Synonyms: NR3C4; KD; AIS; SMAX1; HUMARA; AR
- Molecular weight: 99 kDa (predicted and observed)
- Appearance: Liquid
- Identified uses: Laboratory chemicals, manufacture of substances
- Chemical stability: Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.