Learn More
Medchemexpress LLC Androgen receptor Antibody (YA620) | Protein A affinity purified | 99 kDa | 10 UL

Supplier: Medchemexpress LLC HYP8001410UL
Androgen receptor Antibody (YA620) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to androgen receptor.
- Host: rabbit
- Clonality: recombinant, monoclonal
- Species reactivity: human
- SwissProt ID: P10275
- Gene ID: 367 (NCBI)
- Immunogen: synthetic peptide corresponding to human androgen receptor isoform 3. AA range: 595-644
- Application and dilution ratio: WB (Western Blot): 1:500-1:1000; ICC/IF (Immunocytochemistry/Immunofluorescence): 1:50-1:200
- Sensitivity: endogenous
- Conjugation: non-conjugated
- Modification: unmodified
- Isotype: IgG
- Appearance: liquid
- Formulation: supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% sodium azide
- Storage and stability: stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
- Shipping: shipping with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.