Learn More
Medchemexpress LLC Androgen receptor-IN-6 | 3016437-05-8 | 98.1% | 297.37 g/mol | C16H15N3OS | 10 MG

Supplier: Medchemexpress LLC HY15547810MG
Androgen receptor-IN-6 is a small-molecule androgen receptor inhibitor provided as a characterized research reagent. The compound is described by CAS 3016437-05-8, formula C16H15N3OS, and molecular weight 297.37 g/mol. Supplied as a powder with defined storage recommendations, it is intended for in vitro and preclinical studies of androgen receptor and related nuclear receptor pathways and is not for human use.
- Small-molecule androgen receptor inhibitor for research use.
- Characterized by CAS 3016437-05-8, formula C16H15N3OS, and MW 297.37 g/mol.
- High purity, typically around 98.1%.
- Available in small vial quantities suitable for assay development.
- Powder stability supports long-term storage at -20°C; solvent aliquots require low-temperature storage.
- Intended for in vitro and preclinical studies only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.