Learn More
Abcam Recombinant Human Androgen Receptor protein (His tag), 10UG
Supplier:  Abcam AB8212410UG
Encompass_Preferred
Recombinant Human Androgen Receptor protein (His tag) is a Human Fragment protein, in the 650 to 920 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, EMSA, GSA.
Protein/ Peptide/ Enzyme Synonyms: DHTR, NR3C4, AR, Androgen receptor, Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC9071472