Learn More
Medchemexpress LLC Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- | 115375-60-5 | 99.09% | C22H29F3O5S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY75624
Encompass_Preferred
Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- is a drug intermediate for the synthesis of various active compounds. This product boasts a high purity of 99.09%.
  • Used as a drug intermediate for the synthesis of various active compounds.
  • High purity of 99.09%.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-003-58127