Learn More
Selleck Chemical LLC Anetumab-MMAE-D4006-1MG-5
SDP
Supplier:  Selleck Chemical LLC D40061MG5
Encompass_Preferred
Anetumab-MMAE is an antibody-drug conjugate (ADC) composed of a human anti-mesothelin (MSLN) antibody and the anti-mitotic cytotoxic agent monomethyl auristatin (MMAE)
Catalog No. 50-313-6398