Learn More
Medchemexpress LLC Angelicain (Norcimifugin) | 49624-66-0 | 10 MG

Supplier: Medchemexpress LLC HYN694110MG
Angelicain (Norcimifugin) is a constituent of Cimicifuga foetida with anti-inflammatory activity. It is for research use only.
- Purity: 99.91%
- Appearance: Solid, white to off-white
- Solubility: 100 mg/mL in DMSO (in vitro), or ≥ 2.5 mg/mL in various solvent mixtures (in vivo)
- Storage conditions: -20°C, protected from light. In solvent, -80°C for 6 months; -20°C for 1 month, protected from light
- Biological activity: Cytotoxicity against human HepG2 cells with an IC50 of 5.55 μM
- Classification: Phenols, Monophenols
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.