Learn More
Medchemexpress LLC Angelicain (Norcimifugin) | 49624-66-0 | 5 MG

Supplier: Medchemexpress LLC HYN69415MG
Angelicain (Norcimifugin) is a constituent of Cimicifuga foetida with anti-inflammatory activity. It is for research use only and not sold to patients.
- Purity of 99.91%
- Store at -20°C, protected from light
- For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.