Learn More
Medchemexpress LLC Angiopoietin-related protein 7 (canine), recombinant | >95.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7550710UG
Encompass_Preferred
Recombinant canine angiopoietin-related 7 (ANGPTL7) protein expressed in HEK293 cells and supplied with a C-terminal 6xHis tag. The product corresponds to amino acids Q27-P344, has an approximate molecular weight of ~35 kDa, and a purity of >95% by reducing SDS-PAGE. Supplied as 10 μg; freeze aliquots at -20 °C or -80 °C for long-term storage.
Catalog No. 50-002-91747